Order Entry
Denmark
ContactUsLinkComponent
cis-2-(9-Fluorenylmethoxycarbonylamino)cycloheptanecarboxylic acid
cis-2-(9-Fluorenylmethoxycarbonylamino)cycloheptanecarboxylic acid
Catalog # APOSOR309444-250MG
Supplier:  Apollo Scientific
CAS Number:  
cis-2-(9-Fluorenylmethoxycarbonylamino)cycloheptanecarboxylic acid
Catalog # APOSOR309444-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :379.449 g/mol
Storage Temperature:Ambient
CAS Number:1212250-46-8
Formula:C₂₃H₂₅NO₄
MDL Number:MFCD09881424