cis-1-(9-H-Fluoren-9-ylmethoxycarbonylamino)-indan-2-carboxylic acid
Catalog # APOSOR307652-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:399.439 g/mol
Storage Temperature:Ambient
CAS Number:1175651-61-2
Formula:C₂₅H₂₁NO₄
MDL Number:MFCD09260410