Order Entry
Denmark
ContactUsLinkComponent
cis-2-Amino-2-methyl-cyclohexanecarboxylic acid hydrochloride
Irritant
cis-2-Amino-2-methyl-cyclohexanecarboxylic acid hydrochloride
Catalog # APOSOR307582-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
cis-2-Amino-2-methyl-cyclohexanecarboxylic acid hydrochloride
Catalog # APOSOR307582-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :193.671 g/mol
Melting Pt:220…223 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:202921-88-8
Formula:C₈H₁₅NO₂·ClH
MDL Number:MFCD05863637