Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Denmark
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1H-Imidazole-1-sulphonyl azide hydrochloride
1H-Imidazole-1-sulphonyl azide hydrochloride
1H-Imidazole-1-sulphonyl azide hydrochloride
APOSOR301255-1G
:
Apollo Scientific
:
1H-Imidazole-1-sulphonyl azide hydrochloride
APOSOR301255-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
209.614 g/mol
:
99…104 °C
:
GHS08
:
Refrigerator
:
952234-36-5
:
C₃H₃N₅O₂S·ClH
:
MFCD19705431