Order Entry
Denmark
ContactUsLinkComponent
1-[4-(1-Adamantyl)phenoxy]-3-piperidin-1-ylpropan-2-ol hydrochloride
1-[4-(1-Adamantyl)phenoxy]-3-piperidin-1-ylpropan-2-ol hydrochloride
Catalog # APOSOR22453-1G
Supplier:  Apollo Scientific
CAS Number:  
1-[4-(1-Adamantyl)phenoxy]-3-piperidin-1-ylpropan-2-ol hydrochloride
Catalog # APOSOR22453-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :406.001 g/mol
Melting Pt:249…252 °C
Storage Temperature:Ambient
CAS Number:175136-32-0
Formula:C₂₄H₃₅NO₂·ClH
MDL Number:MFCD00218803