Order Entry
Denmark
ContactUsLinkComponent
[4-(2-Methoxyphenoxy)phenyl]methylamine hydrochloride
Irritant
[4-(2-Methoxyphenoxy)phenyl]methylamine hydrochloride
Catalog # APOSOR12955-100MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
[4-(2-Methoxyphenoxy)phenyl]methylamine hydrochloride
Catalog # APOSOR12955-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :265.735 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:1169974-82-6
Formula:C₁₄H₁₅NO₂·ClH
MDL Number:MFCD03840155