
4-[(Pyridin-2-yl)oxy]benzenesulphonyl chloride hydrochloride 95%
Catalog # APOSOR12616-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
ADR:8,III
:306.165 g/mol
UN:3261
:GHS05
Storage Temperature:Ambient
CAS Number:1170110-04-9
Formula:C₁₁H₈ClNO₃S·ClH
MDL Number:MFCD01862556