6-Azido-6-deoxy-L-galactose ≥98%
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: 6-Azido-6-deoxy-L-fucose
CAS Number:70932-63-7
:
Formula:C₆H₁₁N₃O₅
MDL Number:MFCD27927712
Molecular Weight:205.169 g/mol
Melting Pt:135…138 °C
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together