


Lactophenol-picric acid opløsning, Sigma-Aldrich®
Catalog # 41506-100ML-F
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pakn:100 mL
- Seq:2440
Specifications
About this item
:
ADR:8,II
:GHS08
:GHS05
:GHS07
Storage Temperature:Stue temperatur