Azure II, powder, Sigma-Aldrich®
Catalog # 861065-25G
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:625.68 g/mol
Storage Temperature:Ambient
CAS Number:37247-10-2
Formula:C₁₆H₁₈N₃S·C₁₅H₁₆N₃S·2Cl
MDL Number:MFCD00080716