Order Entry
Denmark
ContactUsLinkComponent
Chromeazurol S (Mordant blue 29), Sigma-Aldrich®
Chromeazurol S (Mordant blue 29), Sigma-Aldrich®
Catalog # 199532-25G
CAS Number:  
undefined
Chromeazurol S (Mordant blue 29), Sigma-Aldrich®
Catalog # 199532-25G
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :605.28 g/mol
Storage Temperature:Ambient
CAS Number:1667-99-8
Formula:C₂₃H₁₆Cl₂O₉S
MDL Number:MFCD00001615