Basic Blue 24 (New Methylene Blue) 90% (dye content), Sigma-Aldrich®
Catalog # 202096-10G
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pk:10 g
- Seq:3236
- Environmentally Preferable:
Specifications
About this item
Synonyms: New Methylene Blue N, zinc chloride double salt, New Methylene Blue N
:
:416.05 g/mol
Storage Temperature:Ambient
CAS Number:6586-05-6
:1934-16-3
Formula:C₁₈H₂₂ClN₃S·0.5 ZnCl₂
MDL Number:MFCD00078135
Melting Pt:>240 °C (dec., lit.)