
Acid blue 80 40% (dye content), Sigma-Aldrich®
Catalog # 210323-100G
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:678.682 g/mol
Melting Pt:>300 °C (lit.)
:GHS07
Storage Temperature:Ambient
CAS Number:4474-24-2
Formula:C₃₂H₂₈N₂Na₂O₈S₂
MDL Number:MFCD00001192