Order Entry
Denmark
ContactUsLinkComponent
Ethyl violet, Sigma-Aldrich®
Ethyl violet, Sigma-Aldrich®
Catalog # 228842-25G
CAS Number:  
undefined
Ethyl violet, Sigma-Aldrich®
Catalog # 228842-25G
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :492.13 g/mol
Storage Temperature:Ambient
CAS Number:2390-59-2
Formula:C₃₁H₄₂ClN₃
MDL Number:MFCD00011765