Tartrazine ≥85% (dye content), Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
CAS Number:1934-21-0
:
Formula:C₁₆H₁₂N₄O₉S₂
MDL Number:MFCD00148908
:534.36 g/mol
:M-Clarity™ quality level = MQ200
Storage Temperature:Room Temperature
Specification Test Results
- Appearance (Colour):Orange
- Appearance (Form):Powder
- Infrared spectrum:Conforms to Structure
- Carbon:30.6 - 36.5 %
- Nitrogen:8.9 - 10.8 %
- Dye Content:≥85
- Solubility (Colour):Yellow to Yellow-Orange
- Solubility (Turbidity)
10 mg/mL, H2O:Clear - E1%
in H2O:435 - 500 - Wavelength:425 - 429 nm
- Recommended Retest Period:4 years
Specification Test Results
Frequently Bought Together