
Ferene disodium salt, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
CAS Number:79551-14-7
:MFCD00040462
Formula:C₁₆H₈N₄Na₂O₈S₂
MDL Number:MFCD00040642
:494.37 g/mol
:M-Clarity™ quality level = MQ200
:GHS07
Storage Temperature:Room Temperature
Specification Test Results
- Appearance (colour):White to dark yellow
- Appearance (form):Powder or crystals
- Solubility (colour):Yellow to very dark yellow and light brown-yellow to very dark brown-yellow
- Solubility (turbidity):Clear to slightly hazy
- Solubility (method):Jun-03
- Water:≤13.0 %
- Carbon content:38.87 % (Theory), anhydrous
- Nitrogen content:11.33 % (Theory), anhydrous
- Infrared spectrum:Conforms to structure
- Bio-tests:Suitable for determination of Fe
- Recommended retest period:60 month
Specification Test Results
Frequently Bought Together