Acridine orange (chlorid salt) solution 10 mg/mL in water, Sigma-Aldrich®
Catalog # A8097-10ML
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pk:10 mL
- Seq:2275
Specifications
About this item
:
:301.81 g/mol (anhy)
Storage Temperature:Ambient
CAS Number:65-61-2
:1704465-79-1
Formula:C₁₇H₁₉N₃·HCl·xH₂O
MDL Number:MFCD00012660