Order Entry
Denmark
ContactUsLinkComponent
 
bisBenzimide H 33258 trihydrochloride (Hoechst 33258) ≥98% (by HPLC, TLC), Sigma-Aldrich®
 
undefined
bisBenzimide H 33258 trihydrochloride (Hoechst 33258) ≥98% (by HPLC, TLC), Sigma-Aldrich®

 

  • Appearance (Form):
    Powder
  • Appearance (Colour):
    Yellow to Dark Yellow
  • Solubility (Solvent):
    Water
  • Solubility (Conc):
    9.80 - 10.20 mg/ml
  • Solubility (Turbidity):
    Clear
  • Solubility (Colour):
    Yellow to Brown
  • NMR (Solvent):
    D2O
  • Identity by NMR:
    Consistent
  • NMR (Residual Solvent 1):
    Methanol
  • NMR (% Solvent 1):
    ≤1.00%
  • NMR (Residual Solvent 2):
    Ethanol
  • NMR (% Solvent 2):
    ≤1.00%
  • NMR (Total % Resid.Solvents):
    ≤1.00%
  • Purity (TLC):
    ≥98.00%
  • Purity (HPLC):
    ≥98.00%

 

Frequently Bought Together