Order Entry
Denmark
ContactUsLinkComponent
Naphthol AS-D chloroacetate solution, Sigma-Aldrich®
Naphthol AS-D chloroacetate solution, Sigma-Aldrich®
Catalog # 911-10ML
CAS Number:  
Naphthol AS-D chloroacetate solution, Sigma-Aldrich®
Catalog # 911-10ML
CAS Number:  

Specifications

  • Pk:
    10 mL
  • Seq:
    2275

Specifications

About this item

 :
 :353.8 g/mol
Storage Temperature:Refrigerator
 :1
CAS Number:35245-26-2
Formula:C₂₀H₁₆ClNO₃