Order Entry
Denmark
ContactUsLinkComponent
Trisodium citrate solution, Sigma-Aldrich®
Trisodium citrate solution, Sigma-Aldrich®
Catalog # 915-50ML
CAS Number:  
Trisodium citrate solution, Sigma-Aldrich®
Catalog # 915-50ML
CAS Number:  

Specifications

  • Pk:
    50 mL
  • Seq:
    2370

Specifications

About this item

 :
 :258.07 g/mol
Storage Temperature:Refrigerator
 :1
CAS Number:68-04-2
Formula:C₆H₅Na₃O₇
MDL Number:MFCD00012462