Naphthol AS-BI Alkaline solution, Sigma-Aldrich®
Catalog # 861-10ML
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pk:10 mL
- Seq:2275
Specifications
About this item
:
ADR:8,III
:GHS05
:GHS07
Storage Temperature:Refrigerator
:1
MDL Number:MFCD00283933