Naphthol AS-BI phosphate solution 12.5 mg/mL in DMF, Sigma-Aldrich®
Katalog # 3864-40ML
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pk:40 mL
- Seq:2360
Specifications
About this item
:
:452.1926 g/mol
ADR:3,III
:M-Clarity™ quality level = MQ500 (more info.)
UN:2265
:GHS02
:GHS07
:GHS08
Storage Temperature:Refrigerator
:1
CAS Number:1919-91-1
Formula:C₁₈H₁₅BrNO₆P
MDL Number:MFCD00004058