Order Entry
Denmark
ContactUsLinkComponent
Isoindolin-1-one
Irritant
Isoindolin-1-one
Certificates
undefined
Irritant
Isoindolin-1-one

Specification Test Results

  • Appearance:
    White to light yellow Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    149 to 153 °C
  • HPLC:
    ≥96.0 %

Specification Test Results

Frequently Bought Together