Meropenem trihydrate
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: (4R,5S,6S)-3-(((3S,5S)-5-(Dimethylcarbamoyl)pyrrolidin-3-yl)thio)-6-((R)-1-hydroxyethyl)-4-methyl-7-oxo-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylic acid trihydrate
CAS Number:119478-56-7
:
Formula:C₁₇H₃₁N₃O₈S
MDL Number:MFCD08600005
Boiling Pt:627 °C
Molecular Weight:437.51 g/mol
Flash Pt:>110 °C
:GHS07
Storage Temperature:Freezer
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Almost white to light yellow Powder or crystals
- Infrared spectrum:Conforms
- Water:≤15 %
- HPLC:≥94 %
- Specific optical rotation:-17° to -21° (20°C, 589 nm) (0.5 % in water)
Specification Test Results
Frequently Bought Together