Thioflavin T stain for amyloid, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
CAS Number:2390-54-7
:
Formula:C₁₇H₁₉ClN₂S
MDL Number:MFCD00011944
:318.86 g/mol
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:219-228-9
Specification Test Results
- Dye content:≥65%
Specification Test Results
Frequently Bought Together