Water baths, insulated stainless steel
: LAUDA
Insulated stainless steel baths which can be used up to 200 °C. The outer jacket is made of powder coated sheet steel. All baths are equipped with a built-in drain tap. These high quality, robust baths are available in a variety of sizes and can be combined with immersion thermostats from Lauda or other manufacturers.
Frequently Bought Together