Order Entry
Denmark
ContactUsLinkComponent
Gas filters, P926
Gas filters, P926
Catalog # PRTWP926BKR
Supplier:  Portwest
undefined
Gas filters, P926
Catalog # PRTWP926BKR
Supplier:  Portwest
Specifications
  • Description:
    ABEK2 Gas filter, P906
  • Pk:
    4
Specifications
About this item
P926 filters are anti-gas and protects against contaminant gases. Equipped with a standard threaded connection conforming to EN 148-1 and can be used on full face masks equipped with the same standard connection.
  • Lightweight and comfortable
  • Individually packed for vending machines
These class 2 filters protects from organic and inorganic acid gases and vapours, sulphur dioxide, ammonia and its derivates where the boiling point is higher than 65 °C.
Certifications: EN 148 -1, EN 14387 Type ABEK Class 2