Order Entry
Denmark
Orders LinkContactUsLinkComponent
Electronic earplug, Peltor™
Electronic earplug, Peltor™
Peltor™ electronic earplug, Green
Catalog # MMMAEEP-100EU
Supplier:   3M
Electronic earplug, Peltor™
Catalog # MMMAEEP-100EU
Supplier:  3M
Electronic earplug, Peltor™
Electronic earplug, Peltor™
Peltor™ electronic earplug, Green
Price
2.510,00 kr
Log in for availability
1 * 1 Par
Product Details & Documents
Peltor™ Electronic Earplugs have an intuitive one-button operation that turns the earplugs on, off, and selects volume settings. They’re powered by Li-ion batteries, which can be charged via a micro B USB charging port on the pocket-sized storage case, incorporating an easily visible charging status indicator.
  • Protect hearing from sudden loud (impulse) noises
  • Enables ambient sounds below 82 dB to still be heard
  • Supports easy communication with those nearby
  • Li-ion batteries provide up to 16 hours of continuous operation
  • Multiple types and sizes of replaceable eartips available to help ensure comfortable fit
  • Small, lightweight design, compatible with most head-borne 3M Personal Safety Equipment products
Peltor™ Electronic Earplugs offer an advanced technology that allows easy communication with people nearby, as well as hearing ambient sounds from the surrounding environment, without compromising hearing protection. These earplugs are simple to use, as a single button turns the earplugs on, off, and selects low, normal, or high-volume settings. Li-ion batteries provide up to 16 hours of continuous operation, while a micro B USB charging port on the pocket-sized storage case makes recharging simple. Battery charge time is approximately 180 minutes, from a computer or wall socket.
The earplug case has a clear lid that makes the charging status indicator easy to see, while a rubber sealing ring around the case helps to protect the earplugs in wet and dirty environments. The case’s waterproof rating complies with IP 54 (with cover and USB port closed), while the signal-to-noise ratio (SNR) meets PPE directive 89/686/EEC. Multiple types and sizes of replaceable eartips are available to help ensure a comfortable fit. The small, lightweight design also supports wearer comfort, as well as enabling use with most head-borne 3M Personal Safety Equipment (PPE) products, such as helmets and face shields.
Specifications
  • Colour:
    Green
  • Description:
    Peltor™ electronic earplug