Analytical standard, Diflufenican, reference material, Fluka™
Diflufenican, Reference material, Neat, Glass bottle, 100 mg
Catalog # HONCR1767-100MG
Supplier: Honeywell Chemicals
Analytical standard, Diflufenican, reference material, Fluka™
Catalog # HONCR1767-100MG
Supplier: Honeywell Chemicals
CAS Number:
Jump to:
Product Details & Documents
Specifications
Specifications
- Pack type:Glass bottle
- Pk:100 mg
- Standards:Reference material
- Description:Diflufenican
- Form:Neat
- Cat. No.:HONCR1767-100MG
- Description:Diflufenican
- CAS No.:83164-33-4
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



