Order Entry
Denmark
ContactUsLinkComponent
Celite® 577, filter aid, fine, calcinated, Fluka™
Health
Celite® 577, filter aid, fine, calcinated, Fluka™
Catalog # 22142-1KG
CAS Number:  
Health
Celite® 577, filter aid, fine, calcinated, Fluka™
Catalog # 22142-1KG
CAS Number:  

Specifications

  • Pack type:
    Drum with inliner
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Composition:
    Al₂O₃, 4.0%
  • Appearance of the substance:
    complying
  • pH (10 % susp., 25°C):
    5.0 - 7.0
  • Loss on ignition (900°C):
    0.4 - 2.0 %

Specification Test Results

About this item

 :
Density:2.3 g/cm³
 :60.08 g/mol
 :GHS08
Storage Temperature:Ambient
CAS Number:61790-53-2
Formula:O₂Si
MDL Number:MFCD00132803