Order Entry
Denmark
ContactUsLinkComponent
 
 
 
HMDS (1,1,1,3,3,3-hexamethyldisilazane) ≥98%, GPR RECTAPUR®
 
undefined
 
 
HMDS (1,1,1,3,3,3-hexamethyldisilazane) ≥98%, GPR RECTAPUR®
 

 

  • Assay:
    98
  • Density (20/4):
    0.774 - 0.775

 

Frequently Bought Together