Order Entry
Denmark
Orders LinkContactUsLinkComponent
Spray bottles

Spray bottles

500 ml, 235 mm, 67 mm, Opaque
Catalog # 215-4364
Supplier:   BURKLE
Spray bottles
Catalog # 215-4364
Supplier:  BURKLE
Spray bottles
Spray bottles

500 ml, 235 mm, 67 mm, Opaque

Price
Inquire For Price
Log in for availability
1 * 1 Items
Product Details & Documents

Basic spray bottles are suitable for occasional spraying in various industries.

  • Spray mist is ideal for spraying various liquids over a large area
  • Pressureless spray bottle is made of HDPE
  • Nozzle functions: Stop and spray

Bottle components in contact with media include PP, PE, TPE, and POM.

Specifications
  • Ext.Ø:
    67 mm
  • Capacity:
    500 ml
  • Colour:
    Opaque
  • Height:
    235 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.