Order Entry
Denmark
ContactUsLinkComponent
Decabromodiphenyl ether [13C12] in n-nonane
Decabromodiphenyl ether [13C12] in n-nonane
Catalog # CAMBEO-5003
CAS Number:  
Decabromodiphenyl ether [13C12] in n-nonane
Catalog # CAMBEO-5003
CAS Number:  

Specifications

  • Pk:
    1,2 mL
  • Seq:
    2080

Specifications

About this item

 :
 :971.1 g/mol
CAS Number:562099-68-7
Formula:*C₁₂Br₁₀O