


Pentachlorophenol [13C6] 100 µg/ml in n-nonane
Catalog # CAMBCLM-661-1.2
Supplier: CAMBRIDGE ISOTOPE LABS
CAS Number:
Specifications
- Pk:1,2 mL
- Seq:2080
Specifications
About this item
:
:272.29 g/mol
:GHS02
:GHS08
:GHS07
CAS Number:85380-74-1
Formula:*C₆Cl₅OH