0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Denmark
ContactUsLinkComponent
Antibodies
Anti-GD3 Monoclonal Antibody
Anti-GD3 Monoclonal Antibody
TCIAA2579-0.2ML
:
TCI
Anti-GD3 Monoclonal Antibody
TCIAA2579-0.2ML
:
TCI
Antigen symbol:
GD3
Clonality:
Monoclonal
Form:
Culture Supernatant
Pk:
0,2 mL