Order Entry
Denmark
ContactUsLinkComponent
 
Hog Acylase (from Kidney)
 :  TCI
 
Hog Acylase (from Kidney)
 :  TCI

 

  • CAS No.:
    9012-37-7
  • Storage conditions:
    Freezer
  • Form:
    White to amber powder to crystalline
  • Enzyme name:
    Acylase

 

Frequently Bought Together