Dissociator with 2 tubes, benchtop, gentleMACS™
Catalog # 431-0272
Supplier: MILTENYI BIOTEC
Specifications
- Description:gentleMACS™ dissociator with 2 tubes
- Pk:1 KIT
Specifications
About this item
gentleMACS™ benchtop dissociator is used for semi-automated dissociation of tissues into single-cell suspensions or thorough homogenates.
- Single sample or two samples in parallel can be processed
- Optimized gentleMACS Programs for a variety of specific applications
- Special protocols for various tissues
- Two types of gentleMACS™ tubes enable time-saving and easy dissociation of tissues in a closed system
The standardised tissue dissociation or homogenization procedures make for reliable and reproducible results.
Programs have been developed for the homogenisation of tissues:
total RNA or mRNA isolation from fresh or frozen samples
protein extraction
determination of bacterial or viral load
mitochondria isolation
Pre-set gentleMACS Octo Dissociator Programs have been developed for easy dissociation of tissues into single-cell suspensions, including:
mouse or human tumor
mouse or rat neonatal heart
neural tissue
mouse spleen, lung, lamina propria, muscle, epidermis, or liver
mouse or human skin and more