Ribbons for label printer BP-4000/BP-4320 printers
Supplier: Brady
Certificates
About this item
Strong environmental resistance against smearing and chemicals.
- Standard performance wax ribbon for paper and temporary labelling
- Reliable, sharp-edge print quality to maximise bar code readability
- Wax/resin ribbon formulation
R7960 series ribbons are printed on the appropriate Brady label, gives good solvent resistance, lower for alcohol, high print quality and excellent smear resistance.
Certifications: RoHS compliant with Directive 2011/65/EU and 2015/863. Part does not contain an SVHC.
Frequently Bought Together