Order Entry
Denmark
ContactUsLinkComponent
Thermal transfer printer ribbon, R6606 series
Thermal transfer printer ribbon, R6606 series
Catalog # BRDY622116
Supplier:  Brady
undefined
Thermal transfer printer ribbon, R6606 series
Catalog # BRDY622116
Supplier:  Brady

Specifications

  • Colour:
    Black
  • Description:
    Thermal transfer ribbon, R6600 series, resin formulation, width 40 mm
  • Length:
    300 m
  • Environmentally Preferable:
  • Pk:
    1
  • For:
    300XPlusII, 360XPlusII, 600XPlusII, BP2461, BP3481, BP6441, i7100, MVP200, MVP300, PR200Plus, PR300Plus, PR600Plus printers

Specifications

About this item

Diesel-resistant thermal transfer ribbon on Brady PermaSleeve ™ and Heatex ™ heat shrink tubing. This print ribbon is highly resistant to chemicals and friction.

Packaging: One print ribbon/box.