
Silver tetrafluoroborate for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:14104-20-2
:
Formula:AgBF₄
MDL Number:MFCD00003408
:194.67 g/mol
Melting Pt:200 °C
EINECS:237-956-5
UN:3260
ADR:8,II
:M-Clarity™ quality level = MQ200
:GHS05
Specification Test Results
- Solubility:(20 °C) soluble
- Melting Point:70 - 73 °C
- Molar Mass:194.67 g/mol
Specification Test Results
Frequently Bought Together