Chloramine-T (sodium salt) trihydrate for synthesis, Sigma-Aldrich®
: Merck
:7080-50-4
:
:CH₃C₆H₄SO₂NClNa·3H₂O
:MFCD00149066
:13,02085
:167…170 °C
:1.43 g/cm³ (20 °C)
:192 °C (closed cup)
:Ambient
:204-854-7
:2928
:6.1 (8),II
:M-Clarity™ quality level = MQ200
:281.69 g/mol
:GHS05
:GHS08
:GHS07
- Solubility:150 g/l (25 °C)
- Molar Mass:281.69 g/mol
- Bulk Density:540 - 680 kg/m3
- Flash Point:192 °C
- pH:8 - 10 (50 g/l, H2O, 20 °C)
- Hazard Class:6.1
- Hazard Class Description:Toxic Materials
Frequently Bought Together