Chloramine-T (sodium salt) trihydrate, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Catalog # 1255.0250
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:250 g
- Seq:3545
Specifications
Specification Test Results
- Assay (as Chlorine):min. 24.5 %
- Insoluble in Ethanol:max. 2.0 %
Specification Test Results
About this item
:
Merck Index:13,02085
Melting Pt:167…170 °C
Density:1.43 g/cm³ (20 °C)
Flash Pt:192 °C (closed cup)
Storage Temperature:Ambient
EINECS:204-854-7
UN:3263
ADR:8,III
:281.69 g/mol
:GHS05
:GHS08
:GHS07
CAS Number:7080-50-4
Formula:CH₃C₆H₄SO₂NClNa·3H₂O
MDL Number:MFCD00149066