1,1-Dimethoxy-2-phenylethane ≥98%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 98% CAS No.: 101-48-4 Molecular Formula: C10H14O2 Formula Weight: 166.22 Flash Point: 83°(181°F) Refractive Index: 1.4940 Boiling Point: 220° Beilstein Reference: 879360 MDL No.: MFCD00008487
Synonyms: Phenylacetaldehyde dimethyl acetal
- CAS Number:101-48-4
- Formula:C₆H₅CH₂CH(OCH₃)₂
- MDL Number:MFCD00008487
- Molecular Weight:166.22 g/mol
- Boiling Pt:220 °C
- Melting Pt:62…64 °C
- Density:1.004
- Flash Pt:83 °C (181 °F)
- Storage Temperature:Ambient
- EINECS:202-945-6
2 Options Available Below
Product Details & Documents
Purity: 98% CAS No.: 101-48-4 Molecular Formula: C10H14O2 Formula Weight: 166.22 Flash Point: 83°(181°F) Refractive Index: 1.4940 Boiling Point: 220° Beilstein Reference: 879360 MDL No.: MFCD00008487
Synonyms: Phenylacetaldehyde dimethyl acetal
- CAS Number:101-48-4
- Formula:C₆H₅CH₂CH(OCH₃)₂
- MDL Number:MFCD00008487
- Molecular Weight:166.22 g/mol
- Boiling Pt:220 °C
- Melting Pt:62…64 °C
- Density:1.004
- Flash Pt:83 °C (181 °F)
- Storage Temperature:Ambient
- EINECS:202-945-6
Documents
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



