Order Entry
Denmark
ContactUsLinkComponent
Hydrobromic acid 5 - 10% in methanol for esterification
Flammable
Corrosive
Irritant
Health
Hydrobromic acid 5 - 10% in methanol for esterification
Supplier:  TCI
Certificates
Flammable
Corrosive
Irritant
Health
Hydrobromic acid 5 - 10% in methanol for esterification
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Light yellow clear liquid
  • Concentration (Neutralization titration):
    5.0 to 10.0 w/w%

Specification Test Results

Frequently Bought Together