Poly(2-methyl-2-oxazoline) Azide Terminated n=∼50, ULTROXA®
: TCI
:1262975-46-1
:
:Refrigerator
- Appearance:White to Orange to Green powder to lump
- Average molecular weight:4,080 to 6,240
- Molecular weight distribution:max. 1.2
Frequently Bought Together