
2,4,8,10-Tetraoxaspiro[5.5]undecane-3,9-diylbis(2-methylpropane-2,1-diyl) bis[3-[3-(tert-butyl)-4-hydroxy-5-methylphenyl]propanoate] ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:90498-90-1
:
Formula:C₄₃H₆₄O₁₀
MDL Number:MFCD09838745
:740.98 g/mol
EINECS:410-730-5
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 95.0 %
- Melting point:124.0 to 128.0 deg-C
Specification Test Results
Frequently Bought Together