
2,4,6-Triisopropylbenzenesulphonyl azide ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:36982-84-0
:
Formula:C₁₅H₂₃N₃O₂S
MDL Number:MFCD00859286
:309.428 g/mol
Melting Pt:43°C
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Elemental analysis (Nitrogen):12.90 to 14.40 %
Specification Test Results
Frequently Bought Together