
alpha,alpha,alpha',alpha'-Tetramethyl-1,3-benzenedipropionitrile ≥98.0% (by GC)
Catalog # TCIAT3421-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:68.0 to 72.0 deg-C
Specification Test Results
About this item
:
:240.35 g/mol
:GHS07
CAS Number:69774-36-3
Formula:C₁₆H₂₀N₂