

2,3,6-Trifluorophenol ≥95.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:113798-74-6
:
Formula:C₆H₃F₃O
MDL Number:MFCD00061214
:148.08 g/mol
UN:1325
ADR:4.1,II
:GHS02
:GHS07
Specification Test Results
- Appearance:White to Orange to Green powder to lump
- Purity (GC):min. 95.0 %
- Purity (Neutralization titration):min. 95.0 %
Specification Test Results
Frequently Bought Together