
7,7,8,8-Tetracyanoquinodimethane ≥99.0% (by HPLC, total nitrogen), purified by sublimation
Supplier: TCI
Certificates
About this item
CAS Number:1518-16-7
:
Formula:C₁₂H₄N₄
MDL Number:MFCD00011664
:204.19 g/mol
Melting Pt:289°C
EINECS:216-174-8
UN:3439
ADR:6.1,III
:GHS06
Specification Test Results
- Appearance:Light yellow to Amber to Dark green powder to crystal
- Purity (HPLC):min. 99.0 area%
- Purity (with Total Nitrogen):min. 99.0 %
Specification Test Results
Frequently Bought Together